Learn More
Invitrogen™ C5AR1 Monoclonal Antibody (S5/1)

Description
Mouse anti Human CD88 antibody, clone S5/1 recognizes the C5a receptor (C5aR) CD88, which is predominantly expressed on cells of the myeloid lineage.
Specifications
Specifications
| Antigen | C5AR1 |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | S5/1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | C5AR1 |
| Gene Accession No. | P21730 |
| Gene Alias | anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.