Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5b-9 Antibody (aE11), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB283850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB283850 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB283850 Supplier Novus Biologicals Supplier No. NBP105120MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C5b-9 Monoclonal antibody specifically detects C5b-9 in Human, Porcine, Bacteria, Equine, Baboon samples. It is validated for Flow Cytometry

Specifications

Antigen C5b-9
Applications Flow Cytometry
Classification Monoclonal
Clone aE11
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex
Host Species Mouse
Immunogen Purified C5b-9
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Pig, Bacteria, Equine, Baboon
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.