Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5L2/GPR77 Antibody (468705), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB369153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369153 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369153 Supplier Novus Biologicals Supplier No. FAB4729A
Add to Cart
Add to Cart

Rat Monoclonal Antibody

C5L2/GPR77 Monoclonal specifically detects C5L2/GPR77 in Mouse samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen C5L2/GPR77
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 468705
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77
Host Species Rat
Immunogen Chinese hamster ovary cell line transfected with mouse C5L2, Met1-Val344, Accession # Q8BW93
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27202
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.