Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5L2/GPR77 Antibody (650918), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB340133 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340133 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340133 Supplier Novus Biologicals Supplier No. FAB10254MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C5L2/GPR77 Monoclonal antibody specifically detects C5L2/GPR77 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen C5L2/GPR77
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 650918
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human C5L2/GPR77, Met1-Val337, Accession # Q9P296
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27202
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.