Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf150 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | C6orf150 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-109-2719
![]() ![]() |
Proteintech Group Inc
264161AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The exact function of C6orf150 remains unknown.Specifications
C6orf150 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8N884 | |
115004 | |
C6orf150 Fusion Protein Ag23455 | |
Primary | |
-20°C | |
CGAS |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C6orf150, cGAS, Cyclic GMP-AMP synthase, h-cGAS, MB21D1 | |
CGAS | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title