Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf58 Antibody (1G1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | C6orf58 |
---|---|
Clone | 1G1 |
Concentration | 1.0 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Applications | Immunocytochemistry, Immunofluorescence |
Description
C6orf58 Monoclonal specifically detects C6orf58 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
C6orf58 | |
1.0 mg/mL | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Zebrafish | |
chromosome 6 open reading frame 58, hypothetical protein LOC352999, MGC72003 | |
C6ORF58 | |
IgG1 | |
Protein A purified |
1G1 | |
Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.02% Sodium Azide | |
352999 | |
GST-C6orf58 fusion protein | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title