Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf58 Antibody (1G1), PerCP, Novus Biologicals™
SDP

Catalog No. NB309221 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309221 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309221 Supplier Novus Biologicals Supplier No. NBP250604PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C6orf58 Monoclonal antibody specifically detects C6orf58 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen C6orf58
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1G1
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias chromosome 6 open reading frame 58, hypothetical protein LOC352999, MGC72003
Host Species Mouse
Immunogen GST-C6orf58 fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 352999
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.