Learn More
Description
Specifications
Specifications
| Antigen | C9orf163 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | chromosome 9 open reading frame 163, hypothetical protein LOC158055, RP11-413M3.11 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human C9orf163 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
