Learn More
Description
Specifications
Specifications
| Antigen | C9orf173 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | chromosome 9 open reading frame 173, FLJ40246, hypothetical protein LOC441476 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human LOC441476 (NP_001243630). Peptide sequence GLRLQTGTPQESLPTYTQTLRELLLEQRPLITADLEVPSPTRYQVPSPSV |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
