Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C9orf61 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | C9orf61 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4999
![]() ![]() |
Proteintech Group Inc
256301AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
C9orf61, also named as FAM189A2 and X123, is predomitly expressed in muscle. It has three isoforms with MW 45-50 kDa, 32-35 kDa and 27 kDa.Specifications
C9orf61 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q15884, Q4FZH1 | |
381217, 9413 | |
C9orf61 Fusion Protein Ag22242 | |
Primary | |
-20°C | |
FAM189A2 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C9orf61, FAM189A2, Protein FAM189A2, Protein X123, RP11 548B3.1, X123 | |
Fam189a2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title