Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf66 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB187971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187971 200 μg
NB187972 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB187971 Supplier Novus Biologicals Supplier No. NBP314648200UL
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C9orf66 Polyclonal antibody specifically detects C9orf66 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen C9orf66
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, pH 7.0 with 0.03% Proclin 300
Gene Alias chromosome 9 open reading frame 66, FLJ31158, RP11-59O6.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human C9orf66 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157983
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.