Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf72 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP247146A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP247146A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP247146A32 Supplier Novus Biologicals Supplier No. NBP247146AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C9orf72 Polyclonal specifically detects C9orf72 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen C9orf72
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunocytochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALSFTD, chromosome 9 open reading frame 72, FTDALS
Gene Symbols C9orf72
Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of the human C9orf72 protein [UniProt Q96LT7].
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 203228
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.