Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1151S1

 View more versions of this product

Catalog No. MS1151S1


Only null left
Add to Cart

Description

Description

As CA125 (Ovarian Cancer Antigen) is a tumour marker usually associated with ovarian epithelial malignancies. Serum assays are widely used to detect this protein in the monitoring of ovarian cancers. However, CA125 antigen may also be detected by immunohistochemistry and expression has been found in neoplasms such as seminal vesicle carcinoma and anaplastic lymphoma. CA125 antigen is not exclusively found in malignant tumours.

Host Species: Mouse

Clone: Ov185:1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer

Positive Control: Ovarian CA

Cellular Localization: Extracellular membrane-associated

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CA 125 (Ovarian Cancer Antigen) Ab-1
Monoclonal
Unconjugated
Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
Ov185:1
Mouse
500 μL
Oncogenesis
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.