Learn More
Epredia™ Lab Vision™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody.
$458.00 - $666.00
Specifications
Antigen | CA 125 (Ovarian Cancer Antigen) Ab-1 |
---|---|
Clone | Ov185:1 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1151S1
|
Epredia™
MS1151S1 |
Concentrated | 500 μL |
Each for $458.00
|
|
||||
MS1151S
|
Epredia™
MS1151S |
Concentrated | 1 mL |
Each for $666.00
|
|
||||
MS1151R7
|
Epredia™
MS1151R7 |
Prediluted | 7 mL |
Each for $523.00
|
|
||||
MS1151S0
|
Epredia™
MS1151S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1151R1
|
Epredia™
MS1151R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
MS1151RQ
|
Epredia™
MS1151RQ |
Ready-to-Use | 12 mL | N/A | N/A | N/A | |||
Description
As CA125 (Ovarian Cancer Antigen) is a tumour marker usually associated with ovarian epithelial malignancies. Serum assays are widely used to detect this protein in the monitoring of ovarian cancers. However, CA125 antigen may also be detected by immunohistochemistry and expression has been found in neoplasms such as seminal vesicle carcinoma and anaplastic lymphoma. CA125 antigen is not exclusively found in malignant tumours.Host Species: Mouse
Clone: Ov185:1
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer
Positive Control: Ovarian CA
Cellular Localization: Extracellular membrane-associated
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CA 125 (Ovarian Cancer Antigen) Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer |
Ov185:1 | |
Monoclonal | |
Mouse | |
Oncogenesis | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.