Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CA12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 151801AP150UL

Description
Carbonic Anhydrase XII is a protein coding gene.Specifications
CA12 | |
Polyclonal | |
Unconjugated | |
CA12 | |
CA XII, CA12, Carbonate dehydratase XII, Carbonic anhydrase 12, carbonic anhydrase XII, CAXII, HsT18816, Tumor antigen HOM RCC 3.1.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363085, 76459, 771 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43570, Q8CI85 | |
Ca12, Car12 | |
CA12 Fusion Protein Ag7107 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction