Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CA12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CA12 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0043
![]() ![]() |
Proteintech Group Inc
151801AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Carbonic Anhydrase XII is a protein coding gene.Specifications
CA12 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43570, Q8CI85 | |
363085, 76459, 771 | |
CA12 Fusion Protein Ag7107 | |
Primary | |
-20°C | |
CA12 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CA XII, CA12, Carbonate dehydratase XII, Carbonic anhydrase 12, carbonic anhydrase XII, CAXII, HsT18816, Tumor antigen HOM RCC 3.1.3 | |
Ca12, Car12 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title