Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA125/MUC16 Antibody (10F10), Novus Biologicals™
SDP

Catalog No. NB298723 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298723 100 μL
NB298722 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298723 Supplier Novus Biologicals Supplier No. NBP326127100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen CA125/MUC16
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 10F10
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
Host Species Rabbit
Immunogen A synthesized peptide derived from Human CA125/MUC16 [UniProt Q8WXI7]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 94025
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.