Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CA125/MUC16 Antibody (10F10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CA125/MUC16 |
---|---|
Clone | 10F10 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
CA125/MUC16 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
A synthesized peptide derived from Human CA125/MUC16 [UniProt Q8WXI7] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10F10 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
94025 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title