Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA125/MUC16 Antibody (986811), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311983 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311983 Supplier Novus Biologicals Supplier No. FAB56091PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CA125/MUC16
Applications Flow Cytometry
Classification Monoclonal
Clone 986811
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CA125/MUC-16, Met13360-Gln14347 (Met13472Thr and Gln13957Lys), Accession # NP_078966.2
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 94025
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.