Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA125/MUC16 Antibody (OTI3C6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N272337A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272337A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272337A647 Supplier Novus Biologicals Supplier No. NBP272337AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Western Blot.

Specifications

Antigen CA125/MUC16
Applications Western Blot
Classification Monoclonal
Clone OTI3C6
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
Gene Symbols MUC16
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 13942-14063 and 21117-21238 of human CA125/MUC16 (NP_078966) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 94025.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.