Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA125/MUC16 Antibody (OTI3C6), FITC, Novus Biologicals™
SDP

Catalog No. NB288225 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288225 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288225 Supplier Novus Biologicals Supplier No. NBP272337F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for Western Blot

Specifications

Antigen CA125/MUC16
Applications Western Blot
Classification Monoclonal
Clone OTI3C6
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 13942-14063 and 21117-21238 of human MUC16 (NP_078966) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 94025
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.