Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA125/MUC16 Antibody (X52), Novus Biologicals™
SDP

Catalog No. NB001357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001357 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001357 Supplier Novus Biologicals Supplier No. NBP2116240.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Western Blot, Sandwich ELISA.

Specifications

Antigen CA125/MUC16
Applications ELISA
Classification Monoclonal
Clone X52
Conjugate Unconjugated
Dilution Sandwich ELISA 1:100-1:2000
Formulation PBS (pH 7.4)
Gene Accession No. Q8WXI7
Gene Alias CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
Gene Symbols MUC16
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with CA125 antigen purified from human ovarian carcinoma (MW > 1 MDa).
Quantity 0.2 mg
Research Discipline Cellular Markers, Extracellular Matrix
Gene ID (Entrez) 94025
Test Specificity Epitope specificity group B (ISOBM class Immunofluorescenceication) similar to M-11
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.