Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CA15-3 Antibody (DBMM1), Novus Biologicals™
SDP

Catalog No. NBP226672 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226672 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226672 Supplier Novus Biologicals Supplier No. NBP226672
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CA15-3 Monoclonal specifically detects CA15-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CA15-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DBMM1
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation Tissue culture supernatant with 0.05% Sodium Azide
Host Species Mouse
Immunogen Human breast cancer cell line ZR-75-1
Purification Method Tissue culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to epithelial membrane antigen (EMA), CA15-3, or polymorphic epithelial mucin. This antibody stains an underglycosylated MUC1 often present on carcinoma cells.
Target Species Human
Content And Storage Store at 4°C.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.