Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CA19-9/Sialyl Lewis a (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody.

$905.00

Specifications

Antigen CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) Ab-3
Clone 116-NS-19-9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1811S1
View Documents
Epredia™
MS1811S1
Concentrated 500 μL
Each for $905.00
Only null left
Add to Cart
 
MS1811S
View Documents
Epredia™
MS1811S
Concentrated 1 mL N/A N/A N/A
MS1811R1
View Documents
Epredia™
MS1811R1
Prediluted 1 mL N/A N/A N/A
Description

Description

Host Species: Mouse

Clone: 116-NS-19-9

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Sialic acid-containing residue of the monosialoganglioside

Immunogen: Human colon carcinoma cell line SW1116

Positive Control: Colon CA

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG2a κ
116-NS-19-9
Monoclonal
Mouse
Oncogenesis
Human colon carcinoma cell line SW1116
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.