Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CABP4 Antibody (OTI9A7), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272071A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272071A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272071A405 Supplier Novus Biologicals Supplier No. NBP272071AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CABP4 Monoclonal specifically detects CABP4 in Human samples. It is validated for Western Blot.

Specifications

Antigen CABP4
Applications Western Blot
Classification Monoclonal
Clone OTI9A7
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CaBP4, calcium binding protein 4, CSNB2Bcalcium-binding protein 4
Gene Symbols CABP4
Host Species Mouse
Immunogen Full length recombinant protein of human CABP4 (NP_660201) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57010.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.