Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CABYR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CABYR |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CABYR Polyclonal specifically detects CABYR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CABYR | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
CABYRa, CABYRc, calcium binding tyrosine-(Y)-phosphorylation regulated, Calcium-binding protein 86, calcium-binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2), Cancer/testis antigen 88, CBP86CABYRc/d, CT88MGC9117, fibrousheathin 2, Fibrousheathin II, fibrousheathin-2, FSP-2calcium binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2), FSP2calcium-binding tyrosine phosphorylation-regulated protein, Testis-specific calcium-binding protein CBP86 | |
CABYR | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
26256 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: YNDVPVTEGVVYIEQLPEQIVIPFTDQVACLKENEQSKENEQSPRVSPKSVVEKTTSGMSKKSVESVKLAQLEENAKYSSVYMEAEATALLSD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title