Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA1G Antibody, Novus Biologicals™
SDP

Catalog No. N180105100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N180105100U 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N180105100U Supplier Novus Biologicals Supplier No. NBP180105100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CACNA1G Polyclonal specifically detects CACNA1G in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CACNA1G
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 2.5 μ/mL
Formulation PBS and 2% Sucrose with no preservative
Gene Accession No. O43497
Gene Alias calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit
Gene Symbols CACNA1G
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CACNA1G (NP_938202). Peptide sequence VSRTHSLPNDSYMCRHGSTAEGPLGHRGWGLPKAQSGSVLSVHSQPADTS.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8913
Reconstitution Centrifuge prior to opening. Reconstitute with sterilized PBS to a final concentration of 1 mg/mL. Vortex followed by Centrifuge again to pellet the solution. (20 μL size only)
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.