Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA1G Antibody, Novus Biologicals™
SDP

Catalog No. NBP145239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145239 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145239 Supplier Novus Biologicals Supplier No. NBP145239
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CACNA1G Polyclonal specifically detects CACNA1G in Human, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.

Specifications

Antigen CACNA1G
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_061496.2
Gene Alias calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit
Gene Symbols CACNA1G
Host Species Goat
Immunogen Peptide with sequence C-AEEISKREDASGQLS corresponding to internal region according to NP_061496.2, NP_938192.1, NP_938194.1, NP_938198.1, NP_938199.1, NP_938200.1, NP_938407.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8913
Test Specificity This antibody is expected to recognize isoform 1 (NP_061496.2), 4 (NP_938199.1), 5 (NP_938194.1), 8 (NP_938198.1), 9 (NP_938192.1), 10 (NP_938200.1) and isoform 15 (NP_938407.1).
Target Species Human, Rat, Bovine, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.