Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA1G Antibody (S178A-9), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP259322FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259322FR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259322FR Supplier Novus Biologicals Supplier No. NBP259322FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CACNA1G Monoclonal specifically detects CACNA1G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CACNA1G
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S178A-9
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit
Gene Symbols CACNA1G
Host Species Mouse
Immunogen Fusion protein amino acids 2052-2172 (cytoplasmic C-terminus) of mouse Cav3.1
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8913
Test Specificity Detects <200kDa. Does not cross-react with Cav3.2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.