Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA1I Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP130674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130674 0.5 mg
NB018497 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP130674 Supplier Novus Biologicals Supplier No. NBP130674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CACNA1I Polyclonal specifically detects CACNA1I in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CACNA1I
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml
Formulation Sterilized water with No Preservative
Gene Accession No. Q9P0X4
Gene Alias Ca(v)3.3, calcium channel, voltage-dependent, T type, alpha 1I subunit, voltage-dependent T-type calcium channel subunit alpha-1I, voltage-dependent, alpha 1I subunit, Voltage-gated calcium channel subunit alpha Cav3.3
Gene Symbols CACNA1I
Host Species Rabbit
Immunogen A synthetic peptide from a cytoplasmic domain of human CACNA1I conjugated to an immunogenic carrier protein was used as the antigen. The peptide is homologous in rat and mouse.
Purification Method Ammonium sulfate precipitation
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8911
Test Specificity Appears to be specific for CACNA1I.
Reconstitution Reconstitute 0.5 mg size in 0.5 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the 0.1 mg size is provided in reconstituted format.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.