Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA1S Antibody (1A), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB300542AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300542AI 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300542AI Supplier Novus Biologicals Supplier No. NB300542AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CACNA1S Monoclonal specifically detects CACNA1S in Human, Mouse, Rat, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CACNA1S
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1A
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CACH1, CACN1, CACNL1A3MHS5, calcium channel, L type, alpha 1 polypeptide, isoform 3 (skeletal muscle, Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle, calcium channel, voltage-dependent, L type, alpha 1S subunit, Cav1.1, CCHL1A3, dihydropyridine receptor, dihydropyridine-sensitive L-type calcium channel alpha-1 subunit, HOKPP, HOKPP1, hypokalemic periodic paralysis), hypoPP, TTPP1, voltage-dependent L-type calcium channel subunit alpha-1S, Voltage-gated calcium channel subunit alpha Cav1.1
Gene Symbols CACNA1S
Host Species Mouse
Immunogen Purified rabbit muscle T-tubule dihydropyridine receptor.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 779
Test Specificity Detects 1,4-dihydropyridine (DHP) receptor alpha-1 subunit in skeletal muscle. The DHP Receptor alpha-1 protein is also known as CACNA1S or Cav1.1 alpha-1 subunit.
Target Species Human, Mouse, Rat, Guinea Pig, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.