Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA2D1 Rabbit anti-Human, DyLight 680, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB045265 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045265 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045265 Supplier Novus Biologicals Supplier No. NBP297587FR

Rabbit Polyclonal Antibody

CACNA2D1 Polyclonal specifically detects CACNA2D1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CACNA2D1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CACNA2, CACNL2Adihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit, calcium channel, L type, alpha 2 polypeptide, calcium channel, voltage-dependent, alpha 2/delta subunit 1, CCHL2A, MHS3, voltage-dependent calcium channel subunit alpha-2/delta-1, Voltage-gated calcium channel subunit alpha-2/delta-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CACNA2D1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 781
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.