Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA2D3 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018482 0.1 mg
NBP130557 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018482 Supplier Novus Biologicals Supplier No. NBP1305570.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CACNA2D3 Polyclonal specifically detects CACNA2D3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CACNA2D3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 1:10-1:500
Formulation Sterilized water with No Preservative
Gene Accession No. Q8IZS8
Gene Alias calcium channel, voltage-dependent, alpha 2/delta subunit 3, voltage-dependent calcium channel subunit alpha-2/delta-3, voltage-dependent, alpha 2/delta 3 subunit
Gene Symbols CACNA2D3
Host Species Rabbit
Immunogen A synthetic peptide from the internal region of human CACNA2D3 conjugated to an immunogenic carrier protein was used as the immunogen.
Molecular Weight of Antigen 123 kDa
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55799
Test Specificity Human CACNA2D3. The antigen is identical in mouse and rat.
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Human, Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.