Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA2D3 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | CACNA2D3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Description
CACNA2D3 Polyclonal specifically detects CACNA2D3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CACNA2D3 | |
Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Sterilized water with No Preservative | |
calcium channel, voltage-dependent, alpha 2/delta subunit 3, voltage-dependent calcium channel subunit alpha-2/delta-3, voltage-dependent, alpha 2/delta 3 subunit | |
CACNA2D3 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
123 kDa |
LYOPH | |
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q8IZS8 | |
55799 | |
A synthetic peptide from the internal region of human CACNA2D3 conjugated to an immunogenic carrier protein was used as the immunogen. | |
Primary | |
Human CACNA2D3. The antigen is identical in mouse and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title