Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CACNA2D4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | CACNA2D4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16899220
![]() |
Novus Biologicals
NBP16899220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP168992
![]() |
Novus Biologicals
NBP168992 |
100 μL |
Each for $487.50
|
|
|||||
Description
CACNA2D4 Polyclonal specifically detects CACNA2D4 in Human samples. It is validated for Western Blot.Specifications
| CACNA2D4 | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| calcium channel, voltage-dependent, alpha 2/delta subunit 4, RCD4, voltage-dependent calcium channel subunit alpha-2/delta-4, voltage-gated calcium channel alpha(2)delta-4 subunit, Voltage-gated calcium channel subunit alpha-2/delta-4 | |
| CACNA2D4 | |
| Primary | |
| 128 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q7Z3S7 | |
| 93589 | |
| IgG | |
| This product is specific to Subunit or Isoform: alpha-2/delta-4. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title