Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB2 Antibody (N8b/1), Biotin, Novus Biologicals™
SDP

Catalog No. NBP147607B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147607B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147607B Supplier Novus Biologicals Supplier No. NBP147607B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CACNB2 Monoclonal specifically detects CACNB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CACNB2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S8b-1
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q8VGC3
Gene Alias CAB2, CACNLB2, Calcium channel voltage-dependent subunit beta 2, calcium channel, voltage-dependent, beta 2 subunit, CAVB2, FLJ23743, Lambert-Eaton myasthenic syndrome antigen B, myasthenic (Lambert-Eaton) syndrome antigen B, MYSB, voltage-dependent L-type calcium channel subunit beta-2
Gene Symbols CACNB2
Host Species Mouse
Immunogen Synthetic peptide amino acids 189-205 of rat CavBeta2
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 783
Test Specificity Detects approx 78 kDa. No cross reactivity against CavB1, CavB3, CavB4.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.