Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB3 Antibody (7D1), Novus Biologicals™
SDP

Catalog No. NB177201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177201 Supplier Novus Biologicals Supplier No. NBP313514
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CACNB3 Monoclonal antibody specifically detects CACNB3 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen CACNB3
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 7D1
Conjugate Unconjugated
Formulation PBS
Gene Alias CAB3, CACNLB3, Calcium channel voltage-dependent subunit beta 3, calcium channel, voltage-dependent, beta 3 subunit, voltage-dependent L-type calcium channel subunit beta-3
Host Species Mouse
Immunogen The immunogen used to generate this antibody corresponds to human CACNB3
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 784
Target Species Human, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.