Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB325319 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325319 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325319 Supplier Novus Biologicals Supplier No. NBP299461JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CACNB3 Polyclonal antibody specifically detects CACNB3 in Human samples. It is validated for Western Blot

Specifications

Antigen CACNB3
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CAB3, CACNLB3, Calcium channel voltage-dependent subunit beta 3, calcium channel, voltage-dependent, beta 3 subunit, voltage-dependent L-type calcium channel subunit beta-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CACNB3.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 784
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.