Learn More
Description
Specifications
Specifications
| Antigen | CACNG7 |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 10-50 ug/ml |
| Formulation | Sterile water with No Preservative |
| Gene Alias | calcium channel, voltage-dependent, gamma subunit 7, Neuronal voltage-gated calcium channel gamma-7 subunit, voltage-dependent calcium channel gamma-7 subunit |
| Gene Symbols | CACNG7 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
