Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cactin Antibody, Novus Biologicals™
SDP

Catalog No. NBP170775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP170775 0.1 mg
NBP170775SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP170775 Supplier Novus Biologicals Supplier No. NBP170775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cactin Polyclonal specifically detects Cactin in Human, Porcine, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Cactin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 5-7 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q8WUQ7
Gene Alias C19orf29, cactin, cactin homolog, cactin, spliceosome C complex subunit, chromosome 19 open reading frame 29, FLJ17482, FLJ59622, fSAPc, functional spliceosome-associated protein c, NY REN 24 antigen, NY-REN-24, NY-REN-24 antigen
Gene Symbols CACTIN
Host Species Rabbit
Immunogen A portion of amino acid 460-510 of human Cactin was used as the immunogen.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58509
Test Specificity Cross reacts with Chimpanzee, Cow, Dog, Human, Opossum, Rhesus Monkey and Wild Pig. Predicted to react with Bacteria, Mosquito, Mouse, Pufferfish, Rat, Wild Turkey, Zebrafish. The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, rhesus monkey, 88% homologous in wild pig, 82% homologous in dog, cow, opossum, 64% homologous in mouse, rat, wild turkey, mosquito and 47% homologous in zebrafish, pufferfish, and bacteria.
Target Species Human, Pig, Bovine, Canine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.