Learn More
Cactin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Cactin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4909
![]() |
Bethyl Laboratories, Inc
A303349AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Cactin is a novel negative regulator of TLR signaling and reveals its capacity to target MHC Class III genes at the molecular and functional level. It is localized to the nucleus, which is critical for manifesting its inhibitory effects on TLR signaling. Experimental evidences show overexpression of Cactin suppresses TLR-induced activation of NF-kB and interferon-regulatory factor transcription factors and induction of TLR-responsive genes. It is also reported Cactin interacts with IkB-like protein and targets other proteins that are encoded by genes in the MHC Class III region of chromosome 6.Specifications
Cactin | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
NY-REN-24, cactin, fSAPc, NY REN 24 antigen, NY-REN-24 antigen, cactin homolog, functional spliceosome-associated protein c, renal carcinoma antigen NY-REN-24, uncharacterized protein C19orf29 | |
CACTIN | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8WUQ7 | |
58509 | |
Between 75 and 125 | |
Primary | |
4° C | |
CACTIN |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.