Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAD Rabbit anti-Human, Mouse, Clone: 0O8D6, Novus Biologicals™
SDP

Catalog No. NB168392 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168392 20 μg
NB168391 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168392 Supplier Novus Biologicals Supplier No. NBP31665020UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CAD Monoclonal antibody specifically detects CAD in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen CAD
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 0O8D6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CAD protein, CAD trifunctional protein, carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, anddihydroorotase, EC 2.1.3.2, EC 3.5.2.3, EC 6.3.5, EC 6.3.5.5, multifunctional protein CAD
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CAD (P27708).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 790
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.