Learn More
CaddyStack™ Electricity Kit
SCI_ED
Catalog No. p-3608000 Shop All Si Metric Manufacturing Limited Products
Click to view available options
Product Type:
Electricity Kit
Teacher'S Guide
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Includes
  • Economy bulb holders (16)
  • Economy switches (16)
  • Miniature bulbs (16)
  • Wire connectors (30)
  • teacher's guide
  • Battery holders (16)
  • D cell batteries (16)
  • Motors (2)
  • Buzzers (2)
Designed with both quality and economy in mind.
  • Complete classroom set of basic electrical components
  • Teacher's guide contains activities for designing and building simple circuits, switches, series and parallel circuits, motors and buzzers
  • Packaged in a CaddyStack
Specifications
Product Type Electricity Kit
For Use With (Application) For designing and building simple circuits, switches, series and parallel circuits, motors and buzzers
Includes 16 economy bulb holders, 16 economy switches, 16 miniature bulbs, 30 wire connectors, teacher's guide, 16 battery holders, 16 d cell batteries, 2 motors, 2 buzzers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.