Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-11 Antibody (667039), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB367639 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367639 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367639 Supplier Novus Biologicals Supplier No. FAB17901Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-11 Monoclonal antibody specifically detects Cadherin-11 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Cadherin-11
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 667039
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, CDHOB, OB, OB-cadherin, Osteoblast cadherin
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cadherin-11, Phe23-Thr617, Accession # AAA35622
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1009
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.