Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-12 Antibody (343621), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB313675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313675 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313675 Supplier Novus Biologicals Supplier No. FAB2240PEATT594

Rat Monoclonal Antibody

Cadherin-12 Monoclonal antibody specifically detects Cadherin-12 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Cadherin-12
Applications Flow Cytometry
Classification Monoclonal
Clone 343621
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias BR-cadherin, cadherin 12, type 2 (N-cadherin 2), cadherin-12, FLJ34857, N-cadherin 2, Neural type cadherin 2, neural type, 2, neuronal cadherin 2
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-12, Met1-Pro609, Accession # P55289
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1010
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.