Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-13 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP22661101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22661101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22661101 Supplier Novus Biologicals Supplier No. NBP2266110.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cadherin-13 Polyclonal specifically detects Cadherin-13 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Cadherin-13
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/ml, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_001248
Gene Alias cadherin 13, H-cadherin (heart), cadherin-13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, Truncated cadherin, truncated-cadherin
Gene Symbols CDH13
Host Species Rabbit
Immunogen T-cadherin antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human T-cadherin.
Molecular Weight of Antigen 80 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1012
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.