Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-16 Antibody (CDH16/2448), HRP, Novus Biologicals™
SDP

Catalog No. NB389032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389032 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389032 Supplier Novus Biologicals Supplier No. NBP308959H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Cadherin-16
Applications ELISA, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone CDH16/2448
Conjugate HRP
Formulation PBS
Gene Alias cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Host Species Mouse
Immunogen Recombinant fragment (around aa 371-507) of human Cadherin-16 protein (exact sequence is proprietary) (Uniprot: O75309)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1014
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.