Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Cadherin-16 Antibody Pair [HRP]
SDP

Catalog No. p-7183060 Shop All Novus Biologicals Products
Click to view available options
Quantity:
15 Plates

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cadherin-16.

Specifications

Assay Range 109.4 pg/mL
Assay Sensitivity 109.38-7000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cadherin-16.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Gene ID (Entrez) 1014
Gene Symbol CDH16
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-Cadherin-16 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.2 mg/mL of mouse anti-Cadherin-16 monoclonal antibody,
  • Standard: Each vial contains 100 ng of recombinant Cadherin-16.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Cadherin-16
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 7 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.