Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-16 Antibody (rCDH16/1071), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB383868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383868 Supplier Novus Biologicals Supplier No. NBP308881G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-16 Monoclonal antibody specifically detects Cadherin-16 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen Cadherin-16
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rCDH16/1071
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Host Species Mouse
Immunogen Recombinant full-length human Cadherin-16 protein (Uniprot: O75309)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1014
Target Species Human, Mouse, Rat, Canine, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.