Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-17, Mouse anti-Human, Clone: CDH17/2618, Novus Biologicals™
SDP

Catalog No. NB010464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010464 20 μg
NB010463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010464 Supplier Novus Biologicals Supplier No. NBP27972320UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-17 Monoclonal specifically detects Cadherin-17 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Cadherin-17
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone CDH17/2618
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q12864
Gene Alias cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024
Gene Symbols CDH17
Host Species Mouse
Immunogen Recombinant fragment (around aa 242-418) of human Cadherin-17 (exact sequence is proprietary)
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1015
Test Specificity This antibody recognizes a protein of 120kDa, which is identified as Cadherin-17 (also known as LI Cadherin).
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.